Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLAIN2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA544942
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA544942 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA544942 Supplier Invitrogen™ Supplier No. PA544942
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: LSAKSGGGPG SGPRRTSSEE LRDATSLLAA GEGGLLDEVE PLRPDELERL Sequence homology: Cow: 92%; Guinea Pig: 86%; Human: 100%; Mouse: 86%; Rabbit: 92%; Rat: 86%.

SLAIN2 is closely related to SLAIN1, a novel stem cell protein that is expressed at the stem cell and epiblast stages. Unlike SLAIN1, SLAIN2 is widely expressed and is thought to play a role in regulating microtubule (MT) growth and organization. SLAIN2 binds to end-binding proteins (EBs), cytoplasmic linker proteins (CLIPs) such as CLIP170, CLIP-associated proteins, and interacts with the MT polymerase ch-TOG. Depletion of SLAIN2 using RNAi led to disorganization of the radial MT array, indicating its importance in the regulation of MT growth and organization.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLAIN2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLAIN2
Gene Accession No. Q9P270
Gene Alias 5033405K12Rik; 8030444K12Rik; AI596370; FLJ21611; hypothetical protein LOC534203; KIAA1458; LOW QUALITY PROTEIN: SLAIN motif-containing protein 2; RGD1311593; SLAIN motif family member 2; SLAIN motif family, member 2; SLAIN motif-containing protein 2; SLAIN2
Gene Symbols SLAIN2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the N-terminal of human SLAIN2 (aa 71-120).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57606
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.