Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLAM/CD150 Antibody (8), PE, Novus Biologicals™

Rabbit Monoclonal Antibody
Supplier: Novus Biologicals NBP289678PE
Description
SLAM/CD150 Monoclonal antibody specifically detects SLAM/CD150 in Human samples. It is validated for ELISA, Immunohistochemistry (Paraffin)Specifications
SLAM/CD150 | |
Monoclonal | |
PE | |
PBS | |
Rabbit | |
Protein A purified | |
RUO | |
Primary | |
Human | |
Purified |
ELISA, Immunohistochemistry (Paraffin) | |
8 | |
ELISA, Immunohistochemistry-Paraffin | |
CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen | |
This antibody was obtained from a rabbit immunized with purified, recombinant Human SLAM/CD150 (Accession#: NP_003028.1; Met 1-Pro 258). | |
0.1 mL | |
Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
6504 | |
Store at 4C in the dark. | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction