Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLAM/CD150 Antibody (MM0553-6L6), DyLight 405, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP211907V
Description
SLAM/CD150 Monoclonal specifically detects SLAM/CD150 in Human samples. It is validated for Western Blot.Specifications
SLAM/CD150 | |
Monoclonal | |
DyLight 405 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
SLAMF1 | |
Human recombinant SLAM/CD150 (extracellular domain) | |
0.1 mL | |
Primary | |
Human | |
Purified |
Western Blot | |
MM0553-6L6 | |
Western Blot | |
CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen | |
Mouse | |
Protein G purified | |
Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
6504 | |
Store at 4C in the dark. | |
IgG2 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction