Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLAM/CD150 Antibody (SR2260), Novus Biologicals™

Rabbit Monoclonal Antibody
$309.50 - $423.00
Specifications
Antigen | SLAM/CD150 |
---|---|
Clone | SR2260 |
Dilution | Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunoprecipitation 1:50 |
Applications | Western Blot, Flow Cytometry, Immunoprecipitation |
Classification | Monoclonal |
Description
SLAM/CD150 Monoclonal antibody specifically detects SLAM/CD150 in Human samples. It is validated for Western Blot, Flow Cytometry, ImmunoprecipitationSpecifications
SLAM/CD150 | |
Western Blot 1:500-1:2000, Flow Cytometry 1:50, Immunoprecipitation 1:50 | |
Monoclonal | |
Purified | |
RUO | |
Human | |
CD 150, CD150IPO-3, CDw150, signaling lymphocytic activation molecule, signaling lymphocytic activation molecule family member 1, SLAMCD150 antigen | |
A synthesized peptide derived from human SLAM/CD150 (Uniprot #: Q13291) | |
Primary | |
Store at -20° C. Avoid freeze/thaw cycles. |
SR2260 | |
Western Blot, Flow Cytometry, Immunoprecipitation | |
Unconjugated | |
Rabbit | |
Cell Cycle and Replication, Hematopoietic Stem Cell Markers, Phospho Specific, Stem Cell Markers | |
PBS, pH 7.4, 150mM NaCl, 50% glycerol. | |
6504 | |
IgG | |
Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title