Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Abnova™ SLAMF1 (Human) ELISA Kit
Shop All Abnova Corporation Products
Click to view available options
Quantity:
1 Kit
Description
- SLAMF1 (Human) ELISA Kit is a sandwich enzyme immunoassay for the quantitative measurement of human SLAMF1.
Specifications
Specifications
| Description | Signaling lymphocytic activation molecule family member 1 |
| Cross Reactivity | Human |
| Product Type | Kit |
| Gene ID (Entrez) | 6504 |
| Gene Symbol | SLAMF1 |
| Quantity | 1 Kit |
| Storage Requirements | Store at 4°C for 6 months. Store at -20°C for 12 months. Avoid multiple freeze-thaw cycles. |
| Target | Human |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction