Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLAMF8 Monoclonal Antibody (250014)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA523947
Description
In direct ELISAs, no cross-reactivity with recombinant human (rh) SLAMF7 or rhSLAMF6 is observed. Reconstitute at 0.5 mg/mL in sterile PBS.
This gene encodes a member of the CD2 family of cell surface proteins involved in lymphocyte activation. These proteins are characterized by Ig domains. This protein is expressed in lymphoid tissues, and studies of a similar protein in mouse suggest that it may function during B cell lineage commitment. The gene is found in a region of chromosome 1 containing many CD2 genes.
Specifications
SLAMF8 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 5% trehalose and No Preservative | |
Q9P0V8 | |
Slamf8 | |
Mouse myeloma cell line NS0-derived recombinant human BLAME/SLAMF8 Ala23-Asp233. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
Flow Cytometry | |
250014 | |
Unconjugated | |
Slamf8 | |
5830408F06Rik; B lymphocyte activator macrophage expressed; BCM-like membrane protein; Blame; B-lymphocyte activator macrophage expressed; CD353; FLJ20442; MGC129578; SBBI42; SLAM family member 8; SLAMF8 | |
Mouse | |
Protein A/G | |
RUO | |
56833 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction