Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLAMF8 Monoclonal Antibody (6A4E2C4)
GREENER_CHOICE

Catalog No. FERMA548766
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA548766 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA548766 Supplier Invitrogen™ Supplier No. MA548766
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

SLAMF8 Monoclonal Antibody for Western Blot, Flow, ELISA

This gene encodes a member of the CD2 family of cell surface proteins involved in lymphocyte activation. These proteins are characterized by Ig domains. This protein is expressed in lymphoid tissues, and studies of a similar protein in mouse suggest that it may function during B cell lineage commitment. The gene is found in a region of chromosome 1 containing many CD2 genes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLAMF8
Applications ELISA, Flow Cytometry, Western Blot, Immunohistochemistry, Western Blot
Classification Monoclonal
Clone 6A4E2C4
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.05% sodium azide
Gene Slamf8
Gene Accession No. Q9P0V8
Gene Alias 5830408F06Rik; B lymphocyte activator macrophage expressed; BCM-like membrane protein; Blame; B-lymphocyte activator macrophage expressed; CD353; FLJ20442; MGC129578; SBBI42; SLAM family member 8; SLAMF8
Gene Symbols Slamf8
Host Species Mouse
Immunogen Purified recombinant fragment of human CD353 (AA: extra 23-233) expressed in E. Coli.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56833
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.