Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLAMF9 Mouse anti-Mouse, BV480, Clone: M349, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 756078
Description
SLAMF9 Monoclonal antibody specifically detects SLAMF9 in Mouse samples. It is validated for Flow cytometry
Specifications
SLAMF9 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
98365 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Flow Cytometry | |
M349 | |
Brilliant Violet 480 | |
CD2F-10; Cd2f10; SLAM family member 9; Slamf9 | |
Mouse SLAMF9 Fusion Protein | |
50 μg | |
Primary | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction