Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLA/LP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA532121
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532121 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532121 Supplier Invitrogen™ Supplier No. PA532121
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Recommended positive controls: A431. Predicted reactivity: Rat (81%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

SLA/LP (SEPSECS) converts O-phosphoseryl-tRNA(Sec) to selenocysteinyl-tRNA(Sec) required for selenoprotein biosynthesis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLA/LP
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Gene SEPSECS
Gene Accession No. Q9HD40
Gene Alias 9130208G10; AA986712; D5Ertd135e; hypothetical protein LOC393123; liver-pancreas antigen; LP; O-phosphoseryl-tRNA(Sec) selenium transferase; PCH2D; Sec synthase; SecS; Selenocysteine synthase; selenocysteinyl-tRNA(Sec) synthase; Sep (O-phosphoserine) tRNA:Sec (selenocysteine) tRNA synthase; SEPSECS; sep-tRNA:Sec-tRNA synthase; SLA; SLA/LP; SLA/LP autoantigen; sla/lpl; SLA-p35; Soluble liver antigen; soluble liver antigen/liver pancreas antigen; soluble liver antigen/liver pancreas antigen, like; Soluble liver antigen/liver pancreas antigen-like; tRNA(Ser/Sec)-associated antigenic protein; TRNP48; UGA suppressor tRNA-associated protein; UGA suppressor tRNA-associated protein homolog; zgc:55980
Gene Symbols SEPSECS
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLA/LP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51091
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.