Learn More
Eisco™ Slate Specimen
SCI_ED
Catalog No. S24797 Shop All Eisco Products
Change view
Click to view available options
Quantity:
1 Ea.
10/Pk.
12/Pk.
Specimen Size:
2 to 3 cm
7 to 8 cm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Specimen Size
S24797 12/Pk. 2 to 3 cm
S24796 1 Ea. 2 to 3 cm
S26601 10/Pk. 2 to 3 cm
S26624 1 Ea. 7 to 8 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. S24797 Supplier Eisco™ Supplier No. ESNG0063PK12
Add to Cart
Add to Cart
Slate Specimen for use in metamorphic classification and identification exercises.
  • For use in metamorphic rock identification exercises due to slate's recognizable platy foliation
  • For use in classroom identification labs and exercises
  • Correlates well to Next Generation Science Standards Evidence Statements
  • Differentiating between slate and shale offers interesting exercise in determining rock physical properties
  • Forms from compression of shale
  • Denser than shale
  • Retains the fissile properties of shale
Specifications
Product Type Metamorphic Rock
For Use With (Application) Metamorphic classification and identification exercises
Packaging Clear labeled and barcoded bag
Quantity 12/Pk.
Specimen Size 2 to 3 cm
Specimen Type Slate