Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLBP Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA578239
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA578239 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA578239 Supplier Invitrogen™ Supplier No. PA578239
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Control: HepG2, HepG2 cytoplasm and membrane fraction extract Predicted Reactivity: Mouse (95%), Rat (93%), Pig (90%), Chimpanzee (100%), Bovine (93%) Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

This gene encodes a protein that binds to the stem-loop structure in replication-dependent histone mRNAs. Histone mRNAs do not contain introns or polyadenylation signals, and are processed by endonucleolytic cleavage. The stem-loop structure is essential for efficient processing but this structure also controls the transport, translation and stability of histone mRNAs. Expression of the protein is regulated during the cell cycle, increasing more than 10-fold during the latter part of G1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLBP
Applications Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.86 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and 0.025% ProClin 300; pH 7
Gene SLBP
Gene Accession No. Q14493
Gene Alias cb157; ele; hairpin binding protein, histone; HBP; histone binding protein; histone RNA hairpin-binding protein; histone stem loop binding protein; histone stem-loop binding protein; histone stem-loop-binding protein; RNA binding protein; sb:cb157; si:dkey-102m7.2; SLBP; Slbp protein; stem-loop (histone) binding protein; stem-loop binding protein; Unknown (protein for MGC:134385)
Gene Symbols SLBP
Host Species Rabbit
Immunogen Recombinant protein encompassing a sequence within the center region of human SLBP. The exact sequence is proprietary.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 7884
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.