Learn More
SLBP Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A303968AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
This gene encodes a protein that binds to the stem-loop structure in replication-dependent histone mRNAs. Histone mRNAs do not contain introns or polyadenylation signals, and are processed by endonucleolytic cleavage. The stem-loop structure is essential for efficient processing but this structure also controls the transport, translation and stability of histone mRNAs. Expression of the protein is regulated during the cell cycle, increasing more than 10-fold during the latter part of G1.Specifications
SLBP | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q14493 | |
SLBP | |
Between 220 and 270 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
SLBP | |
Stem-loop binding protein, HBP | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
7884 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.