Learn More
Invitrogen™ SLC10A2 Polyclonal Antibody

Description
This antibody is predicted to react with rat based on sequence homology. This antibody is tested in Peptide ELISA: antibody detection limit dilution 32,000.
Specifications
Specifications
| Antigen | SLC10A2 |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3 |
| Gene | Slc10a2 |
| Gene Accession No. | P70172 |
| Gene Alias | Apical sodium-dependent bile acid transporter; ASBT; IBAT; ileal apical sodium-dependent bile acid transporter; ileal Na(+)/bile acid cotransporter; ileal sodium/bile acid cotransporter; Ileal sodium-dependent bile acid transporter; ISBAT; ISBT; Na(+)-dependent ileal bile acid transporter; NTCP2; PBAM; Slc10a2; Sodium/taurocholate cotransporting polypeptide, ileal; sodium/taurocholate-cotransporting polypeptide, ileal; solute carrier family 10 (sodium/bile acid cotransporter family), member 2; solute carrier family 10 (sodium/bile acid cotransporter), member 2; solute carrier family 10 member 2; solute carrier family 10, member 2 |
| Gene Symbols | Slc10a2 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.