Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC10A2 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA552108
Description
Immunogen sequence: NKAEIPESKE NGTEPESSFY KANGGFQPDE Highest antigen sequence identity to the following orthologs: Mouse - 47%, Rat - 42%.
This gene encodes a sodium/bile acid cotransporter. This transporter is the primary mechanism for uptake of intestinal bile acids by apical cells in the distal ileum. Bile acids are the catabolic product of cholesterol metabolism, so this protein is also critical for cholesterol homeostasis. Mutations in this gene cause primary bile acid malabsorption (PBAM); mutations in this gene may also be associated with other diseases of the liver and intestines, such as familial hypertriglyceridemia (FHTG).
Specifications
SLC10A2 | |
Polyclonal | |
Unconjugated | |
Slc10a2 | |
Apical sodium-dependent bile acid transporter; ASBT; IBAT; ileal apical sodium-dependent bile acid transporter; ileal Na(+)/bile acid cotransporter; ileal sodium/bile acid cotransporter; Ileal sodium-dependent bile acid transporter; ISBAT; ISBT; Na(+)-dependent ileal bile acid transporter; NTCP2; PBAM; Slc10a2; Sodium/taurocholate cotransporting polypeptide, ileal; sodium/taurocholate-cotransporting polypeptide, ileal; solute carrier family 10 (sodium/bile acid cotransporter family), member 2; solute carrier family 10 (sodium/bile acid cotransporter), member 2; solute carrier family 10 member 2; solute carrier family 10, member 2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
6555 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.2 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12908 | |
Slc10a2 | |
Recombinant protein corresponding to Human SLC10A2. Recombinant protein control fragment (Product #RP-101763). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction