Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC10A3 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
$206.00 - $487.50
Specifications
| Antigen | SLC10A3 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
NBP17931620
![]() |
Novus Biologicals
NBP17931620UL |
20 μL |
Each for $206.00
|
|
|||||
NBP179316
![]() |
Novus Biologicals
NBP179316 |
100 μL |
Each for $487.50
|
|
|||||
Description
SLC10A3 Polyclonal specifically detects SLC10A3 in Human samples. It is validated for Western Blot.Specifications
| SLC10A3 | |
| Polyclonal | |
| Rabbit | |
| NP_062822 | |
| 8273 | |
| Synthetic peptide directed towards the middle region of human SLC10A3The immunogen for this antibody is SLC10A3. Peptide sequence MTFLSTVAATGFLPLSSAIYSRLLSIHETLHVPISKILGTLLFIAIPIAV. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| solute carrier family 10 (sodium/bile acid cotransporter family), member 3 | |
| SLC10A3 | |
| IgG |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title