Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC10A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA555987
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA555987 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA555987 Supplier Invitrogen™ Supplier No. PA555987
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: LPPNCKRTVC LETGSQNVQL CTAILKLAFP PQFIGSMYMF PLLYALFQSA EAGIFVLIYK MYGSEMLHKR DPLDEDEDTD ISYKKLKEEE MADTSYGTVK AENIIMMETA QTS Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 87%.

Transporter for bile acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC10A4
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC10A4
Gene Accession No. Q96EP9
Gene Alias bile acid transporter SLC10A4; E130304D01; Na(+)/bile acid cotransporter 4; P4; SLC10A4; Sodium/bile acid cotransporter 4; solute carrier family 10 (sodium/bile acid cotransporter family), member 4; solute carrier family 10 member 4; solute carrier family 10, member 4
Gene Symbols SLC10A4
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC10A4. Recombinant protein control fragment (Product #RP-90673).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201780
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.