Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC10A4 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104158
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104158 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104158 Supplier Invitrogen™ Supplier No. PA5104158
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC10A4.

Transporter for bile acids.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC10A4
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC10A4
Gene Accession No. Q96EP9
Gene Alias bile acid transporter SLC10A4; E130304D01; Na(+)/bile acid cotransporter 4; P4; SLC10A4; Sodium/bile acid cotransporter 4; solute carrier family 10 (sodium/bile acid cotransporter family), member 4; solute carrier family 10 member 4; solute carrier family 10, member 4
Gene Symbols SLC10A4
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC10A4(Accession Q96EP9), corresponding to amino acid residues L380-M430.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 201780
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.