Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC10A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542415
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542415 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542415 Supplier Invitrogen™ Supplier No. PA542415
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GYSFAKVCTL PLPVCKTVAI ESGMLNSFLA LAVIQLSFPQ SKANLASVAP Sequence homology: Horse: 86%; Human: 100%; Pig: 79%; Rat: 91%.

SLC10A5 is a new member of Solute Carrier Family 10 (SLC10) and the function remains unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC10A5
Applications Immunohistochemistry, Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC10A5
Gene Accession No. Q5PT55
Gene Alias Na(+)/bile acid cotransporter 5; P5; SLC10A5; Sodium/bile acid cotransporter 5; solute carrier family 10 (sodium/bile acid cotransporter family), member 5; solute carrier family 10 member 5; solute carrier family 10, member 5
Gene Symbols SLC10A5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal of human SLC10A5 (aa 351-400).
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 347051
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.