Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC10A7 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5115907
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5115907 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5115907 Supplier Invitrogen™ Supplier No. PA5115907
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total SLC10A7.

SLC10A7 (solute carrier family 10 (sodium/bile acid cotransporter family), member 7), also known as P7, is a 358 amino acid multi-pass membrane protein belonging to the sodium:bile acid symporter family. Existing as seven alternatively spliced isoforms, SLC10A7 is expressed at high levels in liver and lung, moderate levels in placenta, kidney, spleen, and thymus, and low levels in heart, prostate, and testis. A few members of the sodium:bile acid symporter family, such as NTCP (also known as SLC10A1) and Asbt (also known as SLC10A2), are involved in maintaining enterohepatic circulation of bile acids by mediating the first step of active bile transport through membrane barriers of liver and intestine. Other family members, including SLC10A6, play an important role in the cellular delivery of specific prohormones in testis, placenta, adrenal gland and other peripheral tissues. Orphan carriers such as SLC10A7 are uncharacterized and their functions unknown.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC10A7
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide
Gene SLC10A7
Gene Accession No. Q0GE19, Q5PT50, Q5PT53
Gene Alias 2410193C02Rik; C4orf13; Na(+)/bile acid cotransporter 7; P7; PSEC0051; SBF-domain containing protein; SLC10A7; Sodium/bile acid cotransporter 7; solute carrier family 10 (sodium/bile acid cotransporter family), member 7; solute carrier family 10 (sodium/bile acid cotransporter family), member 7-like; solute carrier family 10 member 7; solute carrier family 10, member 7
Gene Symbols SLC10A7
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC10A7(Accession Q0GE19), corresponding to amino acid residues I237-L287.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 291942, 76775, 84068
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.