Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC11A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5112711
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5112711 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5112711 Supplier Invitrogen™ Supplier No. PA5112711
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC11A1 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

NRAMP1, also known as SLC11A1, is a member of the solute carrier family 11 (proton-coupled divalent metal ion transporters) family and encodes a multi-pass membrane protein. The protein functions as a divalent transition metal (iron and manganese) transporter involved in iron metabolism and host resistance to certain pathogens. Mutations in this gene have been associated with susceptibility to infectious diseases such as leprosy and tuberculosis, and inflammatory diseases such as Crohn disease and rheumatoid arthritis. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC11A1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC11A1
Gene Accession No. P49279, P70553
Gene Alias Bcg; host resistance locus Bcg/Ity/Lsh; integral membrane protein; Itg; Ity; Lsh; natural resistance associated macrophage protein; natural resistance associated macrophage protein 1; natural resistance-associated macrophage protein; natural resistance-associated macrophage protein 1; natural resistance-associated macrophages protein 1; NRAMP; NRAMP 1; Nramp1; Slc11a1; solute carrier 11 family member 1; solute carrier family 11 (proton-coupled divalent metal ion transporter), member 1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1; solute carrier family 11 (sodium/phosphate symporters), member 1; solute carrier family 11 A1; solute carrier family 11 member 1; solute carrier family 11 member a1
Gene Symbols SLC11A1
Host Species Rabbit
Immunogen Recombinant Human Natural resistance-associated macrophage protein 1 protein (1-58AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 316519, 6556
Target Species Human, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.