Learn More
Invitrogen™ SLC11A1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5139811
Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: FGQAFYQKTN QAAFNICANS SLHDYAKIFP MNNATVAVDI Highest antigen sequence indentity to the following orthologs: Mouse - 78%, Rat - 80%.
NRAMP1, also known as SLC11A1, is a member of the solute carrier family 11 (proton-coupled divalent metal ion transporters) family and encodes a multi-pass membrane protein. The protein functions as a divalent transition metal (iron and manganese) transporter involved in iron metabolism and host resistance to certain pathogens. Mutations in this gene have been associated with susceptibility to infectious diseases such as leprosy and tuberculosis, and inflammatory diseases such as Crohn disease and rheumatoid arthritis. Alternatively spliced variants that encode different protein isoforms have been described but the full-length nature of only one has been determined.
Specifications
| SLC11A1 | |
| Polyclonal | |
| Unconjugated | |
| SLC11A1 | |
| Bcg; host resistance locus Bcg/Ity/Lsh; integral membrane protein; Itg; Ity; Lsh; natural resistance associated macrophage protein; natural resistance associated macrophage protein 1; natural resistance-associated macrophage protein; natural resistance-associated macrophage protein 1; natural resistance-associated macrophages protein 1; NRAMP; NRAMP 1; Nramp1; Slc11a1; solute carrier 11 family member 1; solute carrier family 11 (proton-coupled divalent metal ion transporter), member 1; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 1; solute carrier family 11 (sodium/phosphate symporters), member 1; solute carrier family 11 A1; solute carrier family 11 member 1; solute carrier family 11 member a1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 6556 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunocytochemistry | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| P49279 | |
| SLC11A1 | |
| Recombinant protein corresponding to Human SLC11A1. Recombinant protein control fragment (Product #RP-108881). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.