Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC11A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA587729
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA587729 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA587729 Supplier Invitrogen™ Supplier No. PA587729
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive Samples: U-87MG, LO2, SW620, NCI-H460, Mouse kidney, Mouse spleen, Mouse testis, Rat kidney, Rat testis; Cellular Location: Cell membrane, Early endosome, Endosome membrane, Mitochondrion outer membrane, Multi-pass membrane protein.

This gene encodes a member of the solute carrier family 11 protein family. The product of this gene transports divalent metals and is involved in iron absorption. Mutations in this gene are associated with hypochromic microcytic anemia with iron overload. A related solute carrier family 11 protein gene is located on chromosome 2. Multiple transcript variants encoding different isoforms have been found for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC11A2
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.41 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.3
Gene Slc11a2
Gene Accession No. O54902, P49281, P49282
Gene Alias AHMIO1; DCT1; Dct-1; Divalent cation transporter 1; divalent metal transporter 1; DMT1; DMT-1; microcytic anemia, viable anaemia; mk; Natural resistance-associated macrophage protein 2; NRAMP 2; Nramp2; OK/SW-cl.20; SLC11A2; solute carrier family 11 (proton-coupled divalent metal ion transporter), member 2; solute carrier family 11 (proton-coupled divalent metal ion transporters), member 2; solute carrier family 11 member 2; Solute carrier family 11 member 2 (natural resistance-associated macrophage protein 2); van
Gene Symbols Slc11a2
Host Species Rabbit
Immunogen Recombinant protein (or fragment).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 18174, 25715, 4891
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.