Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC12A3 Antibody, DyLight 350, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP144270UV
Description
SLC12A3 Polyclonal antibody specifically detects SLC12A3 in Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)Specifications
SLC12A3 | |
Polyclonal | |
Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence, Immunohistochemistry-Paraffin | |
FLJ96318, Na-Cl symporter, NCCT, solute carrier family 12 (sodium/chloride transporters), member 3, solute carrier family 12 member 3, thiazide-sensitive Na-Cl cotransporter, Thiazide-sensitive sodium-chloride cotransporter, TSCNaCl electroneutral thiazide-sensitive cotransporter | |
Produced against a synthetic peptide mapping to a segment of rat NCC (amino acids 74-95), N-terminal | |
0.1 mL | |
ABC Transporters, Cancer, Neuroscience | |
6559 | |
Store at 4°C in the dark. | |
IgG |
Western Blot, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
DyLight 350 | |
50mM Sodium Borate | |
Rabbit | |
Affinity purified | |
RUO | |
Primary | |
Mouse, Rat | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction