Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC12A3, p Thr53 Antibody, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP260775
Description
SLC12A3 Polyclonal specifically detects SLC12A3 in Mouse, Guinea Pig, Hamster samples. It is validated for Western Blot, Immunocytochemistry/Immunofluorescence.Specifications
SLC12A3 (p Thr53) | |
Polyclonal | |
Western Blot 1:100-1:2000, Immunocytochemistry/Immunofluorescence | |
FLJ96318, Na-Cl symporter, NCCT, solute carrier family 12 (sodium/chloride transporters), member 3, solute carrier family 12 member 3, thiazide-sensitive Na-Cl cotransporter, Thiazide-sensitive sodium-chloride cotransporter, TSCNaCl electroneutral thiazide-sensitive cotransporter | |
Rabbit | |
160 kDa | |
100 μL | |
6559 | |
Store at -20C. Avoid freeze-thaw cycles. |
Western Blot, Immunocytochemistry, Immunofluorescence | |
Unconjugated | |
10 mM HEPES (pH 7.5), 150 mM NaCl, 100 ug per ml BSA and 50% glycerol. with No Preservative | |
SLC12A3 | |
Synthetic phospho-peptide corresponding to amino acid residues surrounding Thr53 conjugated to KLH. Accession # P59158 | |
Antigen Affinity-purified | |
Primary | |
Mouse, Guinea Pig, Hamster | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction