Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC12A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA577816
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA577816 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA577816 Supplier Invitrogen™ Supplier No. PA577816
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

1 μg/mL of PA5-77816 was sufficient for detection of NCC3 in 10 μg of rat kidney tissue lysate by colorimetric immunoblot analysis using Goat anti-rabbit IgG:HRP as the secondary antibody. Detects approximately 160kDa.

The SLC12A3 gene encodes the thiazide-sensitive sodium-chloride symporter (NCC), which is critical for salt balance and blood pressure regulation in humans. This symporter is predominantly expressed in the distal convoluted tubule of the kidney, where it facilitates the reabsorption of sodium and chloride ions from the renal filtrate back into the bloodstream. NCC activity plays a vital role in maintaining extracellular fluid volume and potassium homeostasis, impacting systemic blood pressure. Mutations in SLC12A3 can lead to Gitelman syndrome, a hereditary salt-wasting disorder characterized by hypokalemic metabolic alkalosis, low blood pressure, and hypomagnesemia. This gene is regulated by the renin-angiotensin system, which modulates NCC activity through phosphorylation mediated by WNK kinases, thereby affecting renal sodium handling and blood pressure modulation. Research highlights the therapeutic potential of targeting NCC with thiazide diuretics to treat hypertension, emphasizing its importance in both physiological and pathological contexts. The dynamic regulation of NCC by hormonal pathways underscores its significance in fluid and electrolyte balance, contributing to cardiovascular health.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC12A3
Applications Immunohistochemistry, Immunomicroscopy, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.09% sodium azide; pH 7.4
Gene Slc12a3
Gene Accession No. P55017, P55018, P59158
Gene Alias AI035291; chemokine (C-C); CKb10; CK-beta-10; MCP-4; MGC17134; Na-Cl cotransporter; NaCl electroneutral thiazide-sensitive cotransporter; Na-Cl symporter; Ncc; NCC1; NCC-1; NCCT; SCYA13; SCYL1; si:ch211-122c9.4; SLC12A3; sodium chloride cotransporter; solute carrier family 12 (sodium/chloride transporter), member 3; solute carrier family 12 (sodium/chloride transporters), member 3; solute carrier family 12 member 3; solute carrier family 12, member 3; thiazide-sensitive Na-Cl cotransporter; thiazide-sensitive sodium chloride cotransporter; thiazide-sensitive sodium-chloride cotransporter; TSC; wu:fj83c03
Gene Symbols Slc12a3
Host Species Rabbit
Immunogen Produced against a synthetic peptide mapping to a segment of rat NCC (amino acids 74-95), N-terminal.
Purification Method Affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20497, 478118, 54300, 6559
Target Species Human, Mouse, Rat, Canine
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.