Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC12A4 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody
Supplier: Proteintech Group Inc 159271AP150UL

Description
SLC12A4 mediates electroneutral potassium-chloride cotransport when activated by cell swelling. SLC12A4 may contribute to cell volume homeostasis in single cells.Specifications
SLC12A4 | |
Polyclonal | |
Unconjugated | |
SLC12A4 | |
Erythroid K Cl cotransporter 1, hKCC1, KCC1, SLC12A4 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
20498, 29501, 6560 | |
-20°C | |
Liquid |
Immunoprecipitation, Western Blot | |
0.13 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q63632, Q9JIS8, Q9UP95 | |
SLC12A4 | |
SLC12A4 Fusion Protein Ag8762 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction