Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Slc12a5 Mouse anti-Human, Rat, APC, Clone:S1-12, Abnova™

Catalog No. 89333792 Shop All Abnova Corporation Products
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
89-333-792 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 89-333-792 Supplier Abnova Corporation Supplier No. MAB16982
Add to Cart
Add to Cart

Mouse monoclonal antibody raised against partial recombinant rat Slc12a5.

Specifications

Antigen Slc12a5
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Monoclonal
Clone S1-12
Conjugate APC
Description solute carrier family 12 (potassium-chloride transporter), member 5
Dilution Immunocytochemistry (1:100)
Immunofluorescence (1:100)
Immunohistochemistry (Formalin/PFA-fixed paraffin-embedded sections) (1:300)
Immunoprecipitation
Western Blot (1:1000)
The optimal working dilution should be determined by the end user.
Formulation In PBS, pH 7.4 (50% glycerol, 0.09% sodium azide)
Gene Slc12a5
Gene Alias Kcc2
Host Species Mouse
Immunogen Recombinant protein corresponding to amino acids 932-1043 of rat Slc12a5.
Quantity 100 μg
Gene ID (Entrez) 171373
Target Species Human, Rat
Content And Storage Store at -20°C.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.