Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC12A8 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA5107021

Catalog No. PIPA5107021


Only null left
Add to Cart

Description

Description

Sequence homology: Cow: 100%; Dog: 100%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

SLC12A8 is a cation/chloride cotransporter that may play a role in the control of keratinocyte proliferation.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC12A8
Polyclonal
Unconjugated
SLC12A8
Cation-chloride cotransporter 9; CCC9; E330020C02Rik; SLC12A8; solute carrier family 12 (potassium/chloride transporters), member 8; solute carrier family 12 (sodium/potassium/chloride transporters), member 8; solute carrier family 12 member 8
Rabbit
Affinity Chromatography
RUO
84561
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Western Blot, Surface Plasmon Resonance
0.5 mg/mL
PBS with 2% sucrose and 0.09% sodium azide; pH 7.4
A0AV02
SLC12A8
Synthetic peptide directed towards the N terminal region of human SLC12A8.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.