Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC12A9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA584762
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA584762 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA584762 Supplier Invitrogen™ Supplier No. PA584762
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: AWHSARLRIF LCLGPREAPG AAEGRLRALL SQLRIRAEVQ EVVWGEGAGA GEPEAEEEGD FVNSGRGDAE AEALAR Highest antigen sequence indentity to the following orthologs: Rat - 93%, Mouse - 92%.

SLC12A9 is a protein coding gene. May be an inhibitor of SLC12A1. Seems to correspond to a subunit of a multimeric transport system and thus, additional subunits may be required for its function.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC12A9
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.3 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC12A9
Gene Accession No. Q9BXP2
Gene Alias Cation-chloride cotransporter 6; cation-chloride cotransporter-interacting protein 1; Ccc6; CCC-interacting protein 1; CIP1; hCCC6; Potassium-chloride transporter 9; Slc12a8; SLC12A9; solute carrier family 12 (potassium/chloride transporters), member 9; solute carrier family 12 member 9; solute carrier family 12, member 9; WO3.3
Gene Symbols SLC12A9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC12A9. Recombinant protein control fragment (Product #RP-108530).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 56996
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.