Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC13A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553262
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553262 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553262 Supplier Invitrogen™ Supplier No. PA553262
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ATSAMMVPIA HAVLDQLHSS QASSNVEEGS NNPTFELQEP SPQKEVTKLD NGQALPVTSA SSEGRAHLSQ KHLHLTQCMS LCVCYSASIG GIATLTGTAP NLVLQGQINS LFPQNGNVV Highest antigen sequence identity to the following orthologs: Mouse - 67%, Rat - 71%.

SLC13A2 belongs to the SLC13A transporter (TC 2.A.47) family, NADC subfamily. It is a multi-pass membrane protein. SLC13A2 cotransports of sodium ions and dicarboxylates such as succinate and citrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC13A2
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC13A2
Gene Accession No. Q13183
Gene Alias hypothetical protein LOC505069; INDY; intestinal sodium/dicarboxylate cotransporter; mNaDC-1; mucin; Na(+)/dicarboxylate cotransporter 1; Na(+)-coupled citrate transporter; NaCT; Nadc1; naDC-1; Renal sodium/dicarboxylate cotransporter; SDCT1; Slc13a2; sodium/dicarboxylate cotransporter; sodium/dicarboxylate co-transporter; sodium-dependent dicarboxylate transporter; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 2; solute carrier family 13 member 2; solute carrier family 13, member 2
Gene Symbols SLC13A2
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC13A2. Recombinant protein control fragment (Product #RP-91205).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9058
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.