Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC13A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5121202
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5121202 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. PIPA5121202 Supplier Invitrogen™ Supplier No. PA5121202
Add to Cart
Add to Cart
This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: 293T, U-937, MCF7, Mouse kidney, Mouse liver, Rat kidney. The target is usually found in the following locations: Membrane, Multi-pass membrane protein. Immunogen sequence: TREPGFFLGW GNLAFPNAKG ESMVSDGTVA IFIGIIMFII PSKFPGLTQD PENPGKLKAP LGLLDWKTVN QKMPWNIVLL LGGGYALAKG SERSGLSEWL GNKLTPLQSV P.

SLC13A2 belongs to the SLC13A transporter (TC 2.A.47) family, NADC subfamily. It is a multi-pass membrane protein. SLC13A2 cotransports of sodium ions and dicarboxylates such as succinate and citrate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC13A2
Applications ELISA, Western Blot
Classification Polyclonal
Concentration 0.939 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene SLC13A2
Gene Accession No. P70545, Q13183, Q9ES88
Gene Alias hypothetical protein LOC505069; INDY; intestinal sodium/dicarboxylate cotransporter; mNaDC-1; mucin; Na(+)/dicarboxylate cotransporter 1; Na(+)-coupled citrate transporter; NaCT; Nadc1; naDC-1; Renal sodium/dicarboxylate cotransporter; SDCT1; Slc13a2; sodium/dicarboxylate cotransporter; sodium/dicarboxylate co-transporter; sodium-dependent dicarboxylate transporter; solute carrier family 13 (sodium-dependent dicarboxylate transporter), member 2; solute carrier family 13 member 2; solute carrier family 13, member 2
Gene Symbols SLC13A2
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 345-455 of human SLC13A2 (NP_003975.1).
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 20500, 65202, 9058
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.