Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC13A5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542526
Description
Peptide sequence: CKAMTLCICY AASIGGTATL TGTGPNVVLL GQMNELFPDS KDLVNFASWF Sequence homology: Cow: 92%; Dog: 85%; Guinea Pig: 100%; Horse: 85%; Human: 100%; Mouse: 77%; Rabbit: 100%; Rat: 93%.
S13A5 is a tricarboxylate plasma transporter with a preference for citrate.
Specifications
SLC13A5 | |
Polyclonal | |
Unconjugated | |
SLC13A5 | |
EIEE25; Indy; mINDY; Na(+)/citrate cotransporter; Na+-coupled citrate transporter protein; NaC2/NaCT; NACT; novel solute carrier family 13 (sodium-dependent dicarboxylate transporter) (Slc13a2 or 3) member; SLC13A5; Sodium-coupled citrate transporter; sodium-dependent citrate transporter; sodium-dependent dicarboxylate transporter; solute carrier family 13 (sodium-dependent citrate transporter), member 5; solute carrier family 13 member 5 | |
Rabbit | |
Affinity Chromatography | |
RUO | |
284111 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q86YT5 | |
SLC13A5 | |
Synthetic peptide directed towards the middle region of human SLC13A5. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction