Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC13A5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5113058
Description
SLC13A5 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA
S13A5 is a tricarboxylate plasma transporter with a preference for citrate.
Specifications
| SLC13A5 | |
| Polyclonal | |
| Unconjugated | |
| SLC13A5 | |
| EIEE25; Indy; mINDY; Na(+)/citrate cotransporter; Na+-coupled citrate transporter protein; NaC2/NaCT; NACT; novel solute carrier family 13 (sodium-dependent dicarboxylate transporter) (Slc13a2 or 3) member; SLC13A5; Sodium-coupled citrate transporter; sodium-dependent citrate transporter; sodium-dependent dicarboxylate transporter; solute carrier family 13 (sodium-dependent citrate transporter), member 5; solute carrier family 13 member 5 | |
| Rabbit | |
| Protein G | |
| RUO | |
| 237831, 266998, 284111 | |
| -20°C or -80°C if preferred | |
| Liquid |
| ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4 | |
| Q67BT3, Q86YT5, Q8CJ44 | |
| SLC13A5 | |
| Recombinant Human Solute carrier family 13 member 5 protein (156-204AA). | |
| 100 μg | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction