Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
SLC14A1 Antibody (888418) [Biotin], Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals FAB8238B
Description
SLC14A1 Monoclonal antibody specifically detects SLC14A1 in Human samples. It is validated for Flow Cytometry, CyTOFSpecifications
SLC14A1 | |
Monoclonal | |
Biotin | |
PBS | |
Mouse | |
Protein A or G purified from hybridoma culture supernatant | |
RUO | |
6563 | |
Store at 4°C in the dark. | |
IgG2b |
Flow Cytometry, CyTOF | |
888418 | |
Flow Cytometry, CyTOF-ready | |
FLJ33745, HsT1341, HUT11, JKFLJ41687, RACH1, solute carrier family 14 (urea transporter), member 1 (Kidd blood group), Solute carrier family 14 member 1, truncated urea transporter, urea transporter 1, urea transporter JK glycoprotein, Urea transporter, erythrocyte, urea transporter-B1, UT1UT-B1, UTEblood group Kidd urea transporter | |
NS0 mouse myeloma cell line transfected with human SLC14A1, Accession # Q13336 | |
0.1 mL | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction