Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC14A1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA563749
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA563749 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA563749 Supplier Invitrogen™ Supplier No. PA563749
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: GHYNPFFPAK LVIPITTAPN ISWSDLSALE LLKSIPVGVG QI Highest antigen sequence identity to the following orthologs: Mouse - 74%, Rat - 74%.

SLC14A1 is a specialized low-affinity urea transporter. It mediates urea transport in erythrocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC14A1
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC14A1
Gene Accession No. Q13336
Gene Alias 2610507K20Rik; 3021401A05Rik; HsT1341; HUT11; JK; Kidd blood group; null; RACH1; RACH2; SLC14A1; SLC14A1 JK; solute carrier family 14 (urea transporter), member 1; solute carrier family 14 (urea transporter), member 1 (Kidd blood group); solute carrier family 14 member 1; solute carrier family 14 member 1 (Kidd blood group); urea transporter; urea transporter 1; Urea transporter B; urea transporter JK glycoprotein; urea transporter variant 2; Urea transporter, erythrocyte; urea transporter-B1; UrT2; UT1; UT-1; UT11; UT3; UT-B; Utb1; UT-B1; UTE
Gene Symbols SLC14A1
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC14A1. Recombinant protein control fragment (Product #RP-101940).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6563
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.