Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC14A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5110377
Description
Immunogen sequence: FYLQAKKRMV ESPL.
SLC14A1 is a specialized low-affinity urea transporter. It mediates urea transport in erythrocytes.
Specifications
SLC14A1 | |
Polyclonal | |
Unconjugated | |
SLC14A1 | |
2610507K20Rik; 3021401A05Rik; HsT1341; HUT11; JK; Kidd blood group; null; RACH1; RACH2; SLC14A1; SLC14A1 JK; solute carrier family 14 (urea transporter), member 1; solute carrier family 14 (urea transporter), member 1 (Kidd blood group); solute carrier family 14 member 1; solute carrier family 14 member 1 (Kidd blood group); urea transporter; urea transporter 1; Urea transporter B; urea transporter JK glycoprotein; urea transporter variant 2; Urea transporter, erythrocyte; urea transporter-B1; UrT2; UT1; UT-1; UT11; UT3; UT-B; Utb1; UT-B1; UTE | |
Rabbit | |
Affinity Chromatography | |
RUO | |
108052, 54301, 6563 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
2.1 mg/mL | |
PBS with 50% glycerol and 0.05% ProClin 300; pH 7.3 | |
P97689, Q13336, Q8VHL0 | |
SLC14A1 | |
A synthetic peptide corresponding to a sequence within amino acids 158-257 of human SLC14A1 (NP_0012952081). | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction