Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC14A1 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA543133
Description
Peptide sequence: LSSPLMCLHA AIGSLLGIAA GLSLSAPFEN IYFGLWGFNS SLACIAMGGM Sequence homology: Cow: 86%; Dog: 93%; Goat: 93%; Horse: 83%; Human: 100%; Mouse: 83%; Pig: 93%; Sheep: 93%.
SLC14A1 is a specialized low-affinity urea transporter. It mediates urea transport in erythrocytes.
Specifications
SLC14A1 | |
Polyclonal | |
Unconjugated | |
SLC14A1 | |
2610507K20Rik; 3021401A05Rik; HsT1341; HUT11; JK; Kidd blood group; null; RACH1; RACH2; SLC14A1; SLC14A1 JK; solute carrier family 14 (urea transporter), member 1; solute carrier family 14 (urea transporter), member 1 (Kidd blood group); solute carrier family 14 member 1; solute carrier family 14 member 1 (Kidd blood group); urea transporter; urea transporter 1; Urea transporter B; urea transporter JK glycoprotein; urea transporter variant 2; Urea transporter, erythrocyte; urea transporter-B1; UrT2; UT1; UT-1; UT11; UT3; UT-B; Utb1; UT-B1; UTE | |
Rabbit | |
Affinity Chromatography | |
RUO | |
6563 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Immunohistochemistry, Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q13336 | |
SLC14A1 | |
Synthetic peptide directed towards the C-terminal of human SLC14A1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction