Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC14A2/UT-A Rabbit anti-Human, Polyclonal, Bioss
SDP

Catalog No. 501981679
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-198-1679 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-198-1679 Supplier Bioss Supplier No. BS11801R
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Water balance is tightly regulated in the kidneys. Through urinary concentration mechanisms, the kidney prevents the unnecessary excrement of water, thereby protecting the body from dehydration. UT-A (Urea transporter, kidney), also known as SLC14A2 and HUT2, is a 920 amino acid multi-pass membrane protein that functions as a specialized low-affinity vasopressin-regulated urea-transporter. Expressed in the inner medulla of the kidney and localized to the apical membrane, UT-A mediates rapid transepithelial urea transport across the inner medullary collecting duct. There are two isoforms of UT-A that are produced as a result of alternative splicing events. Isoform 1 interacts with Snapin, a SNARE-associated protein, which is thought to help in its recruitment to the plasma membrane. Interestingly, isoform 1 expression is induced by low protein in the diet, whereas isoform 2 expression is induced by dehydration.

Specifications

Antigen SLC14A2/UT-A
Applications Immunofluorescence, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 μg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.09% sodium azide; pH
Gene SLC14A2
Gene Accession No. Q15849
Gene Symbols SLC14A2
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide derived from human SLC14A2/UT-A.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 8170
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.