Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC15A2 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA542480
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA542480 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA542480 Supplier Invitrogen™ Supplier No. PA542480
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GNENNSLLIE SIKSFQKTPH YSKLHLKTKS QDFHFHLKYH NLSLYTEHSV Sequence homology: Cow: 86%; Dog: 83%; Guinea Pig: 79%; Horse: 93%; Human: 100%; Mouse: 79%; Pig: 86%; Rabbit: 93%; Rat: 79%.

SLC15A2 belongs to the PTR2/POT transporter (TC 2.A.17) family. It is a multi-pass membrane protein. The expression/activity of PEPT2 (SLC15A2) may be a critical factor in the modulation of opioidergic neurotransmission in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC15A2
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Slc15a2
Gene Accession No. Q16348
Gene Alias 8430408C16Rik; C78862; H+ coupled di/tri-peptide cotransporter 2; kidney H(+)/peptide cotransporter; oligopeptide transporter; oligopeptide transporter, kidney isoform; OPT-3; Pept2; Peptide transporter 2; proton-dependent high affinity oligopeptide transporter PepT2; SLC15A2; Slc15a2 variant A; Slc15a2 variant A2; solute carrier family 15 (H+/peptide transporter), member 2; solute carrier family 15 (oligopeptide transporter), member 2; solute carrier family 15 member 2; solute carrier family 15, member 2
Gene Symbols Slc15a2
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human SLC15A2.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 6565
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.