Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC15A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA548477
Change view
Click to view available options
Quantity:
400 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA548477 400 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA548477 Supplier Invitrogen™ Supplier No. PA548477
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Predicted to react with rat based on sequence homology.

Proton oligopeptide cotransporter. Transports free histidine and certain di- and tripeptides. Target protein may play a role in protein catabolism in the lymphatic system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC15A3
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene SLC15A3
Gene Accession No. Q8IY34
Gene Alias cAMP inducible gene 1; cAMP-inducible gene 1 protein; Ci1; cI-1; OCTP; Osteoclast transporter; peptide transporter 3; Peptide/histidine transporter 2; PHT2; PTR3; Slc15a3; solute carrier family 15 (oligopeptide transporter), member 3; solute carrier family 15 member 3; solute carrier family 15, member 3
Gene Symbols SLC15A3
Host Species Rabbit
Immunogen KLH conjugated synthetic peptide between 168-197 amino acids from the Central region of human SLC15A3.
Purification Method Antigen affinity chromatography, Protein A
Quantity 400 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51296
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.