Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC15A3 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5113780
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5113780 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5113780 Supplier Invitrogen™ Supplier No. PA5113780
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: ILPVMVTLVP YWMVYFQMQS TYVLQGLHLH IPNIFRTNPN ISLLLRSDSS For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 93%; Dog: 93%; Guinea Pig: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Rabbit: 100%; Rat: 100%.

Proton oligopeptide cotransporter. Transports free histidine and certain di- and tripeptides. Target protein may play a role in protein catabolism in the lymphatic system.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC15A3
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene SLC15A3
Gene Accession No. Q8BPX9
Gene Alias cAMP inducible gene 1; cAMP-inducible gene 1 protein; Ci1; cI-1; OCTP; Osteoclast transporter; peptide transporter 3; Peptide/histidine transporter 2; PHT2; PTR3; Slc15a3; solute carrier family 15 (oligopeptide transporter), member 3; solute carrier family 15 member 3; solute carrier family 15, member 3
Gene Symbols SLC15A3
Host Species Rabbit
Immunogen Synthetic peptide directed towards the C-terminal region of Mouse Slc15a3 (aa 314-363).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 65221
Target Species Mouse
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.