Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

SLC15A5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA559657
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA559657 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA559657 Supplier Thermo Scientific Supplier No. PA559657
Add to Cart
Add to Cart

SLC15A5 Polyclonal Antibody

SLC15A5 Polyclonal Antibody for IHC

TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC15A5
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC15A5
Gene Accession No. A6NIM6
Gene Alias Peptide/histidine transporter ENSP00000340402; SLC15A5; Solute carrier family 15 member 5; solute carrier family 15, member 5
Gene Symbols SLC15A5
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC15A5. Recombinant protein control fragment (Product #RP-97880).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 729025
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.