Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC16A10 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA548538
Description
SLC16A10 Polyclonal Antibody for Western Blot, IHC (P), Flow
Sodium-independent transporter that mediates the update of aromatic acid. Can function as a net efflux pathway for aromatic amino acids in the basosolateral epithelial cells.
Specifications
SLC16A10 | |
Polyclonal | |
Unconjugated | |
SLC16A10 | |
2610103N14Rik; 9830169E08; aromatic amino acid transporter 1; MCT 10; MCT10; monocarboxylate transporter; Monocarboxylate transporter 10; PRO0813; Slc16a10; solute carrier family 16 (aromatic amino acid transporter), member 10; solute carrier family 16 (monocarboxylic acid transporters), member 10; solute carrier family 16 member 10; solute carrier family 16, member 10 (aromatic amino acid transporter); TAT1; T-type amino acid transporter 1 | |
Rabbit | |
Antigen affinity chromatography, Protein A | |
RUO | |
117247 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Western Blot | |
0.4 mg/mL | |
PBS with 0.09% sodium azide; pH 7.4 | |
Q8TF71 | |
SLC16A10 | |
KLH conjugated synthetic peptide between 301-329 amino acids from the C-terminal region of human SLC16A10. | |
400 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction