Learn More
Invitrogen™ SLC16A10 Polyclonal Antibody

Description
The antibody detects endogenous levels of total SLC16A10 protein.
Specifications
Specifications
| Antigen | SLC16A10 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 1.2 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 |
| Gene | SLC16A10 |
| Gene Accession No. | Q3U9N9, Q8TF71 |
| Gene Alias | 2610103N14Rik; 9830169E08; aromatic amino acid transporter 1; MCT 10; MCT10; monocarboxylate transporter; Monocarboxylate transporter 10; PRO0813; Slc16a10; solute carrier family 16 (aromatic amino acid transporter), member 10; solute carrier family 16 (monocarboxylic acid transporters), member 10; solute carrier family 16 member 10; solute carrier family 16, member 10 (aromatic amino acid transporter); TAT1; T-type amino acid transporter 1 |
| Gene Symbols | SLC16A10 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.