Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC16A10 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553507
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553507 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553507 Supplier Invitrogen™ Supplier No. PA553507
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: SKKQREISKT TGKEKMEKML ENQNSLLSSS SGMFKKESDS I Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 88%.

Sodium-independent transporter that mediates the update of aromatic acid. Can function as a net efflux pathway for aromatic amino acids in the basosolateral epithelial cells.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC16A10
Applications Immunohistochemistry (Paraffin), Immunocytochemistry
Classification Polyclonal
Concentration 0.1 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene SLC16A10
Gene Accession No. Q8TF71
Gene Alias 2610103N14Rik; 9830169E08; aromatic amino acid transporter 1; MCT 10; MCT10; monocarboxylate transporter; Monocarboxylate transporter 10; PRO0813; Slc16a10; solute carrier family 16 (aromatic amino acid transporter), member 10; solute carrier family 16 (monocarboxylic acid transporters), member 10; solute carrier family 16 member 10; solute carrier family 16, member 10 (aromatic amino acid transporter); TAT1; T-type amino acid transporter 1
Gene Symbols SLC16A10
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human SLC16A10. Recombinant protein control fragment (Product #RP-91140).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 117247
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.