Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC16A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598710
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598710 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598710 Supplier Invitrogen™ Supplier No. PA598710
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC16A11 Polyclonal Antibody for Western Blot, ICC/IF, ELISA

Proton-linked monocarboxylate transporter. It catalyzes the transport of pyruvate across the plasma membrane (PubMed:28666119). Probably involved in hepatic lipid metabolism: overexpression results in an increase of triacylglycerol(TAG) levels, small increases in intracellular diacylglycerols and decreases in lysophosphatidylcholine, cholesterol ester and sphingomyelin lipids (PubMed:24390345). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC16A11
Applications ELISA, Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 3.78 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene SLC16A11
Gene Accession No. Q8NCK7
Gene Alias AB041591; MCT 11; MCT11; MNCb-2717; monocarboxylate transporter; Monocarboxylate transporter 11; monocarboxylic acid transporter 11; monocarboxylic acid transporters; novel RNA polymerase Rpb1 C-terminal repeat domain containing protein; Slc16a11; solute carrier family 16 (monocarboxylic acid transporters), member 11; solute carrier family 16 member 11; solute carrier family 16, member 11; solute carrier family 16, member 11 (monocarboxylic acid transporter 11)
Gene Symbols SLC16A11
Host Species Rabbit
Immunogen Recombinant Human Monocarboxylate transporter 11 protein (428-471AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162515
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.