Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC16A11 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5103924
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5103924 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5103924 Supplier Invitrogen™ Supplier No. PA5103924
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total MOT11.

Proton-linked monocarboxylate transporter. It catalyzes the transport of pyruvate across the plasma membrane (PubMed:28666119). Probably involved in hepatic lipid metabolism: overexpression results in an increase of triacylglycerol(TAG) levels, small increases in intracellular diacylglycerols and decreases in lysophosphatidylcholine, cholesterol ester and sphingomyelin lipids (PubMed:24390345). [UniProt].
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC16A11
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene SLC16A11
Gene Accession No. Q5NC32, Q8NCK7
Gene Alias AB041591; MCT 11; MCT11; MNCb-2717; monocarboxylate transporter; Monocarboxylate transporter 11; monocarboxylic acid transporter 11; monocarboxylic acid transporters; novel RNA polymerase Rpb1 C-terminal repeat domain containing protein; Slc16a11; solute carrier family 16 (monocarboxylic acid transporters), member 11; solute carrier family 16 member 11; solute carrier family 16, member 11; solute carrier family 16, member 11 (monocarboxylic acid transporter 11)
Gene Symbols SLC16A11
Host Species Rabbit
Immunogen A synthesized peptide derived from human SLC16A11(Accession Q8NCK7), corresponding to amino acid residues M1-G31.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 162515, 216867, 287450
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.