Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC16A11 Polyclonal Antibody

Rabbit Polyclonal Antibody

Supplier:  Invitrogen™ PA524397

Catalog No. PIPA524397


Only null left
Add to Cart

Description

Description

SLC16A11 Polyclonal Antibody for Western Blot, IHC (P), Flow

Proton-linked monocarboxylate transporter. It catalyzes the transport of pyruvate across the plasma membrane (PubMed:28666119). Probably involved in hepatic lipid metabolism: overexpression results in an increase of triacylglycerol(TAG) levels, small increases in intracellular diacylglycerols and decreases in lysophosphatidylcholine, cholesterol ester and sphingomyelin lipids (PubMed:24390345). [UniProt].
TRUSTED_SUSTAINABILITY
Specifications

Specifications

SLC16A11
Polyclonal
Unconjugated
SLC16A11
AB041591; MCT 11; MCT11; MNCb-2717; monocarboxylate transporter; Monocarboxylate transporter 11; monocarboxylic acid transporter 11; monocarboxylic acid transporters; novel RNA polymerase Rpb1 C-terminal repeat domain containing protein; Slc16a11; solute carrier family 16 (monocarboxylic acid transporters), member 11; solute carrier family 16 member 11; solute carrier family 16, member 11; solute carrier family 16, member 11 (monocarboxylic acid transporter 11)
Rabbit
Antigen affinity chromatography, Protein A
RUO
162515, 216867
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot
0.48 mg/mL
PBS with 0.09% sodium azide; pH 7.4
Q5NC32, Q8NCK7
SLC16A11
KLH conjugated synthetic peptide between 48-76 amino acids from the N-terminal region of human SLC16A11.
400 μL
Primary
Human, Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Product Certifications
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.