Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ SLC16A12 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA542536
Description
Peptide sequence: WMIVAGCFLV TICTRAVTRC ISIFFVEFQT YFTQDYAQTA WIHSIVDCVT Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 100%; Human: 100%; Mouse: 86%; Rabbit: 93%; Rat: 93%.
This gene encodes a transmembrane transporter that likely plays a role in monocarboxylic acid transport. A mutation in this gene has been associated with juvenile cataracts with microcornea and renal glucosuria.
Specifications
SLC16A12 | |
Polyclonal | |
Unconjugated | |
Slc16a12 | |
3230401A21; AW210596; CJMG; Creatine transporter 2; CRT2; LOW QUALITY PROTEIN: monocarboxylate transporter 12; MCT 12; MCT12; monocarboxylate transporter; Monocarboxylate transporter 12; monocarboxylic acid transporter 12; MOT12; RGD1311468; SLC16A12; solute carrier family 16 (monocarboxylic acid transporters), member 12; solute carrier family 16 member 12; solute carrier family 16, member 12; solute carrier family 16, member 12 (monocarboxylic acid transporter 12) | |
Rabbit | |
Affinity chromatography | |
RUO | |
387700 | |
-20°C, Avoid Freeze/Thaw Cycles | |
Liquid |
Western Blot | |
0.5 mg/mL | |
PBS with 2% sucrose and 0.09% sodium azide | |
Q6ZSM3 | |
Slc16a12 | |
Synthetic peptide directed towards the N-terminal of human SLC16A12 (aa 49-98). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction