Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ SLC16A12 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA598535
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA598535 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA598535 Supplier Invitrogen™ Supplier No. PA598535
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

SLC16A12 Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

This gene encodes a transmembrane transporter that likely plays a role in monocarboxylic acid transport. A mutation in this gene has been associated with juvenile cataracts with microcornea and renal glucosuria.
TRUSTED_SUSTAINABILITY

Specifications

Antigen SLC16A12
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 2.82 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene Slc16a12
Gene Accession No. Q6ZSM3
Gene Alias 3230401A21; AW210596; CJMG; Creatine transporter 2; CRT2; LOW QUALITY PROTEIN: monocarboxylate transporter 12; MCT 12; MCT12; monocarboxylate transporter; Monocarboxylate transporter 12; monocarboxylic acid transporter 12; MOT12; RGD1311468; SLC16A12; solute carrier family 16 (monocarboxylic acid transporters), member 12; solute carrier family 16 member 12; solute carrier family 16, member 12; solute carrier family 16, member 12 (monocarboxylic acid transporter 12)
Gene Symbols Slc16a12
Host Species Rabbit
Immunogen Recombinant Human Monocarboxylate transporter 12 protein (431-486AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 387700
Target Species Human
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.